Learn More
GRIN1 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 27676-1-AP-20UL
Description
The GRIN1 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human GRIN1. This antibody recognizes human, mouse, rat antigen. The GRIN1 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
| GRIN1 | |
| Polyclonal | |
| Unconjugated | |
| 20 μL | |
| 2902 | |
| IgG |
| Western Blot, Immunohistochemistry, ELISA | |
| 200 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Rat, Mouse |