missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GRIA1 (phospho S845) Rabbit anti-Human, Mouse, Rat, Clone:RM296, Abnova™
Rabbit monoclonal antibody raised against human GRIA1 (phospho S845).
Brand: Abnova MAB16921.100uL
This item is not returnable.
View return policy
Description
Glutamate receptors are the predominant excitatory neurotransmitter receptors in the mammalian brain and are activated in a variety of normal neurophysiologic processes. These receptors are heteromeric protein complexes with multiple subunits, each possessing transmembrane regions, and all arranged to form a ligand-gated ion channel. The classification of glutamate receptors is based on their activation by different pharmacologic agonists. This gene belongs to a family of alpha-amino-3-hydroxy-5-methyl-4-isoxazole propionate (AMPA) receptors. Alternatively spliced transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]Specifications
| GRIA1 (phospho S845) | |
| Monoclonal | |
| Unconjugated | |
| Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:500-1000) Western Blot (1:1000-2000) The optimal working dilution should be determined by the end user. |
|
| GRIA1 | |
| Rabbit | |
| 100 μL | |
| This antibody reacts to human Glutamate Receptor 1, AMPA Receptor (GluR1) only when phosphorylated at Ser845. There is no cross-reactivity to GluR1 that is not phosphorylated at Ser845. This antibody may also react to mouse or rat Phospho-AMPA Receptor (GluR1) (Ser845) as predicted by immunogen homology. | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| RM296 | |
| glutamate receptor, ionotropic, AMPA 1 | |
| In PBS (50% glycerol, 1% BSA, 0.09% sodium azide) | |
| GLUH1, GLUR1, GLURA, HBGR1, MGC133252 | |
| A phospho-peptide corresponding to human GRIA1 (phospho S845). | |
| 2890 | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction