missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gremlin 1 Rabbit anti-Mouse, Clone: 117, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90353-50UL
This item is not returnable.
View return policy
Beschreibung
Gremlin 1 Monoclonal specifically detects Gremlin 1 in Mouse samples. It is validated for ELISA.
Spezifikation
| Gremlin 1 | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| ELISA | |
| 117 | |
| ELISA 1:5000-1:10000 | |
| Cell proliferation-inducing gene 2 protein, CKTSF1B1gremlin 1, cysteine knot superfamily, homolog, Cysteine knot superfamily 1, BMP antagonist 1gremlin 1, cysteine knot superfamily, homolog (Xenopus laevis), DAN domain family member 2, DAND2GREMLIN, Down-regulated in Mos-transformed cells protein, DRMMGC126660, gremlin, gremlin 1, gremlin 1-like protein, gremlin-1, IHG-2, Increased in high glucose protein 2, increased in high glucose-2, proliferation-inducing gene 2 | |
| Recombinant Mouse Gremlin 1 protein | |
| 50 μg | |
| Neuroscience | |
| 26585 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur