missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ASAHL/N-acylethanolamine-hydrolyzing Acid Amidase Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-62534
This item is not returnable.
View return policy
Description
ASAHL/N-acylethanolamine-hydrolyzing Acid Amidase Polyclonal specifically detects ASAHL/N-acylethanolamine-hydrolyzing Acid Amidase in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| ASAHL/N-acylethanolamine-hydrolyzing Acid Amidase | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| Acid ceramidase-like protein, ASAHL, ASAH-like protein, EC 3.5.1.-, N-acylethanolamine acid amidase, N-acylethanolamine-hydrolyzing acid amidase, N-acylsphingosine amidohydrolase (acid ceramidase)-like, N-acylsphingosine amidohydrolase-like, PLT | |
| Rabbit | |
| 37 kDa | |
| 100 μL | |
| Primary | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| Q02083 | |
| NAAA | |
| Synthetic peptides corresponding to ASAHL The peptide sequence was selected from the middle region of ASAHL. Peptide sequence VSWLIRATLSESENFEAAVGKLAKTPLIADVYYIVGGTSPREGVVITRNR. | |
| Affinity purified | |
| RUO | |
| 27163 | |
| Human, Mouse, Rat, Pig, Bovine, Equine, Guinea Pig, Rabbit | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction