Learn More
GRB7 Antibody (OTI15E7), Novus Biologicals Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-44931-30ul
This item is not returnable.
View return policy
Description
GRB7 Monoclonal antibody specifically detects GRB7 in Human,Mouse,Rat samples. It is validated for Western Blot
Specifications
| GRB7 | |
| Monoclonal | |
| Unconjugated | |
| PBS (pH 7.3) 1% BSA, 50% glycerol | |
| Mouse | |
| >95% | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI15E7 | |
| Western Blot 1:500-2000 | |
| B47, Epidermal growth factor receptor GRB-7, GRB7 adapter protein, growth factor receptor-bound protein 7 | |
| Full length human recombinant protein of human GRB7 (NP_005301) produced in E.coli. | |
| 30 μL | |
| Breast Cancer, Cancer, Extracellular Matrix | |
| 2886 | |
| Store at -20 degrees C. Avoid freeze/thaw cycles. | |
| Purified |