Learn More
IBA Lifesciences Strep-Tactin™XT Superflow™ Column
Product Code. 15573077 Shop All IBA Lifesciences Products
Click to view available options
Capacity (Metric):
1 x 1 mL
1 x 5 mL
5 x 0.2 mL
1 x 10 mL
Quantity:
1 Column
5 Columns
Unit Size:
Each
Pack of 5
Product Code. 15573077
Brand: IBA Lifesciences 24014001
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Strep-Tactin™XT Superflow™ column is a gravity flow column to purify Strep-tag™II or Twin-Strep-tag™ fusion proteins
  • Strep-Tactin™XT Superflow™ columns contain a 6% agarose coupled with the streptavidin variant Strep-Tactin™XT and are applicable for purification of Strep-tag™II or Twin-Strep-tag™ fusion proteins via gravity flow.
  • As with any other Strep-Tactin™XT resins, the unspecific protein binding is extremely low, which leads to an outstanding purity of the target protein.
  • In comparison to Strep-Tactin™ Superflow™, Strep-Tactin™XT Superflow™ has a higher affinity for Strep-tag™ fusion proteins, which leads to an improved immobilization and purification.
  • In addition, Strep-Tactin™XT Superflow™ allows the application of denaturing agents such as urea during the purification procedure.
  • To reduce the waiting period during the sample and buffer application, the use of the application aid WET FRED is recommended. 
Specifications
Capacity (Metric) 1 x 10 mL
Quantity 1 Column
For Use With (Application) Protein Purification
Type Gravity Flow Column
For Research Use Only.