Learn More
IBA Lifesciences Gravity flow Strep-Tactin™ 4Flow™ high capacity mini- column
Shop All IBA Lifesciences Products
Click to view available options
Quantity:
0.2 mL
Unit Size:
0.2mL
Description
- Strep-Tactin™ 4Flow™ high capacity columns are designed for purifying Strep-tag™II and Twin-Strep-tag™ proteins via affinity chromatography in gravity flow
- Recommended for all protein classes due to the broad pH compatibility, excellent pressure stability, outstanding target purity (>95%), and high binding affinity
- 4% crosslinked agarose beads coupled to Strep-Tactin™ for straightforward purification of proteins with Strep-tag™II or a Twin-Strep-tag™
- Targeted elution: no tedious titration steps required and multiple regeneration cycles possible using 100 mM NaOH
- Ideal alternative for proteins that are difficult to elute from Strep-Tactin™XT 4Flow™ resins
- Biotinylated proteins also bind to Strep-Tactin™ 4Flow™ high capacity and need to be blocked using BioLock
Specifications
Specifications
| Binding Capacity | 20 mg/mL resin |
| Capacity (Metric) | 0.2 mL |
| Description | 4% agarose, crosslinked |
| Particle Size | 50 to 150 μm |
| Form | Resin Slurry |
| Quantity | 0.2 mL |
| Protein Form | Tetrameric protein |
| Target | Proteins tagged with Twin-Strep-tag™; Strep-tag™II; cross reactivity: biotin |
| Type | Gravity Flow Column |
| Final Product Type | Affinity chromatography equipment |
| Show More |