Learn More
Granulin Antibody (CL5695) [Janelia Fluor™ 669], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-61425JF669
Product Code. 30292518
  • 737.00€ / 0.10mL
Description
Granulin Monoclonal antibody specifically detects Granulin in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Gene Knock-Out
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Granulin
Monoclonal
Janelia Fluor 669
acrogranin, GEP, GP88, granulin, granulin-epithelin, granulins, PC cell-derived growth factor, PCDGF, PEPI, PGRN, proepithelin, progranulin
This antibody was developed using a recombinant protein derived from P28799, with the exact immunogen sequence remaining proprietary.
0.1 mL
Cancer, Cell Cycle and Replication, Lipid and Metabolism
2896
Store at 4⪚C in the dark.
IgG1
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Gene Knock-Out
CL5695
50mM Sodium Borate
Mouse
Protein A purified