missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Granulin Antibody (CL5695) [Alexa Fluor™ 594], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-61425AF594
This item is not returnable.
View return policy
Description
Granulin Monoclonal antibody specifically detects Granulin in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Gene Knock-Out
Specifications
| Granulin | |
| Monoclonal | |
| Alexa Fluor 594 | |
| acrogranin, GEP, GP88, granulin, granulin-epithelin, granulins, PC cell-derived growth factor, PCDGF, PEPI, PGRN, proepithelin, progranulin | |
| This antibody was developed using a recombinant protein derived from P28799, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Cancer, Cell Cycle and Replication, Lipid and Metabolism | |
| 2896 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Gene Knock-Out | |
| CL5695 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction