Learn More
Hirschmann™ Graduated Pipettes with Piston
Product Code. 10014740
Click to view available options
Volume (Metric):
1 mL
2 mL
5 mL
10 mL
20 mL
25 mL
Length (Metric):
335 mm
345 mm
360 mm
385 mm
430 mm
475 mm
Unit Size:
Each
Product Code. 10014740
Brand: Hirschmann™ 1190165
Only null left
Add to basket
Add to basket
Provides volumetric accuracy for your pipetting needs. Hirschmann Instruments™ Graduated Pipettes with Piston come with amber stain graduation.
  • Graduated to tip
  • Available in a range of capacities
Specifications
Graduated Yes
Graduation Color Amber
Graduations 0.1 mL
Graduation Type Amber graduations permanently with silver diffusion color
Quantity 1 Each
Volume (Metric) 20 mL
Length (Metric) 430 mm
Type Graduated Pipet