Learn More
GPX7 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13501-1-AP-20UL
This item is not returnable.
View return policy
Description
Human GPX 7 is believed to be a member of the glutathione peroxidase family of proteins. These proteins play important roles in cellular defense mechanisms against oxidative damage. This is accomplished by catalyzing the reduction of hydroperoxides and hydrogen peroxide through the reducing substrate, glutathione.Specifications
| GPX7 | |
| Polyclonal | |
| Unconjugated | |
| GPX7 | |
| CL683, glutathione peroxidase 7, GPx 7, GPX6, GPX7, GSHPx 7 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 2882 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.31 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q96SL4 | |
| GPX7 | |
| GPX7 Fusion Protein Ag4328 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |