missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GPR85 Monoclonal Antibody (1042202)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA560907
This item is not returnable.
View return policy
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
GPR85, a member of the SREB (Super-Conserved Receptor Expressed in Brain) family of GPCRs, is an Orphan-A GPCR with an unknown ligand. The rat counterpart of SREB2 is involved in the development of the cerebral cortex. GPR85 has been reported to be expressed in several areas of human and rat CNS (Matsumoto et al., 2000), as well as in genital organs. ESTs have been isolated from B-cell/lung/testis, brain, testis, and embryo libraries, as well as many mouse tissue libraries.
Specifications
| GPR85 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| P60893 | |
| GPR85 | |
| Human GPR85 extracellular domain synthetic peptide. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Immunocytochemistry | |
| 1042202 | |
| Unconjugated | |
| GPR85 | |
| 2900026B03Rik; FLJ50993; FLJ51159; FLJ99433; G protein-coupled receptor 85; Gpr85; PKrCx1; probable G-protein coupled receptor 85; Rgpr85; seven transmembrane helix receptor; SREB; SREB2; Srep2; Super conserved receptor expressed in brain 2 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 54329 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction