missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPR37 Antibody (436923), DyLight 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB4450K
This item is not returnable.
View return policy
Description
GPR37 Monoclonal antibody specifically detects GPR37 in Human samples. It is validated for Flow Cytometry (Intracellular Staining), CyTOF
Specifications
| GPR37 | |
| Monoclonal | |
| DyLight 488 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry (Intracellular Staining), CyTOF | |
| 436923 | |
| Intracellular Staining by Flow Cytometry, CyTOF-ready | |
| EDNRBL, Endothelin B receptor-like protein 1, G protein-coupled receptor 37 (endothelin receptor type B-like), hET(B)R-LP, PAELRETBR-LP-1, Parkin-associated endothelin receptor-like receptor, probable G-protein coupled receptor 37 | |
| NS0 mouse myeloma cell line transfected with human GPR37, Ala27-Cys613, Accession # O15354 | |
| 0.1 mL | |
| GPCR | |
| 2861 | |
| Store at 4°C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction