missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPR17 Antibody (CL10474) [DyLight 650], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-07993C
This item is not returnable.
View return policy
Description
GPR17 Monoclonal antibody specifically detects GPR17 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications
| GPR17 | |
| Monoclonal | |
| DyLight 650 | |
| DKFZp686M18273, G protein-coupled receptor 17, G-protein coupled receptor 17, P2Y-like receptor, R12, UDP/CysLT receptor, uracil nucleotide/cysteinyl leukotriene receptor | |
| This antibody was developed using a synthetic peptide derived from Q13304, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| GPCR, Neuroscience | |
| 2840 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| CL10474 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction