missing translation for 'onlineSavingsMsg'
Learn More

GPR153 Antibody - BSA Free, Novus Biologicals™

Product Code. 18389784 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mL
Unit Size:
0.05mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18389784 0.05 mL 0.05mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18389784 Supplier Novus Biologicals Supplier No. NLS2032

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

GPR153 Polyclonal specifically detects GPR153 in Human, Mouse, Rat, Porcine, Bovine, Canine, Hamster, Monkey, Rabbit samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPR153
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin 2 - 12 ug/ml
Formulation PBS with 0.1% Sodium Azide
Gene Alias DKFZp762B2210, G protein-coupled receptor 153, PGR1G-protein coupled receptor PGR1, probable G-protein coupled receptor 153
Gene Symbols GPR153
Host Species Rabbit
Immunogen Synthetic 17 amino acid peptide from 2nd extracellular domain of human GPR153.
Purification Method Affinity Purified
Quantity 0.05 mL
Regulatory Status RUO
Research Discipline GPCR
Primary or Secondary Primary
Gene ID (Entrez) 387509
Test Specificity G Protein-Coupled Receptor LS19072
Target Species Human, Mouse, Rat, Pig, Bovine, Canine, Hamster, Monkey, Rabbit
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.