missing translation for 'onlineSavingsMsg'
Learn More

GPR139 Antibody - BSA Free, Novus Biologicals™

Product Code. 18558399 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mL
Unit Size:
50µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18558399 0.05 mL 50µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18558399 Supplier Novus Biologicals Supplier No. NLS2716

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

GPR139 Polyclonal antibody specifically detects GPR139 in Human, Mouse, Rat, Bovine, Canine, Chicken, Equine, Primate, Monkey, Rabbit samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPR139
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin 6 - 14 μg/mL
Formulation PBS
Gene Alias G protein-coupled receptor 139, G(q)-coupled orphan receptor GPRg1, GPRG1, G-protein-coupled receptor PGR3, PGR3G protein-coupled receptor PGR3, probable G-protein coupled receptor 139
Host Species Rabbit
Immunogen Synthetic 16 amino acid peptide from 3rd cytoplasmic domain of human GPR139.
Purification Method Immunogen affinity purified
Quantity 0.05 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 124274
Target Species Human, Mouse, Rat, Bovine, Canine
Content And Storage Keep as concentrated solution. Aliquot and store at -20°C or below. Avoid multiple freeze/thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.