missing translation for 'onlineSavingsMsg'
Learn More

GPR137 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

415.00€ - 624.00€

Specifications

Antigen GPR137
Applications Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18649515
View Documents
Novus Biologicals
NBP2-38976-25ul
25 μL
415.00€
25µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18176418
View Documents
Novus Biologicals
NBP2-38976
0.1 mL
624.00€
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

GPR137 Polyclonal specifically detects GPR137 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications

Specifications

GPR137
Polyclonal
Rabbit
GPCR
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
C11orf4, C11orf4GPR137ATM7SF1L1Transmembrane 7 superfamily member 1-like 1 protein, G protein-coupled receptor 137, GPR137A, integral membrane protein GPR137, TM7SF1L1
GPR137
IgG
Affinity Purified
Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
RUO
Human
Q96N19
56834
This antibody was developed against a recombinant protein corresponding to amino acids: LLAQDPGGRGYPLLWPAPCCSCHSELVP
Primary
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.