missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPR107 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
292.00€
Specifications
| Antigen | GPR107 |
|---|---|
| Dilution | Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500-1:1000 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18438040
|
Novus Biologicals
NBP1-83629-25ul |
25 μL |
292.00€
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18246198
|
Novus Biologicals
NBP1-83629 |
0.1 mL |
N/A
|
N/A | |||||
Description
GPR107 Polyclonal specifically detects GPR107 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| GPR107 | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 57720 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:KFRPASDNPYLQLSQEEEDLEMESVVTTSGVMESMKKVKKVTNGSVEPQGEWEGSV | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500-1:1000 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| bA138E2.2, DKFZp667C222, FLJ16312, FLJ20998, FLJ22591, G protein-coupled receptor 107, KIAA1624GCDRP, Lung seven transmembrane receptor 1, LUSTR1MGC126118, MGC15440, protein GPR107, RP11-88G17 | |
| GPR107 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title