missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPD2 Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 68174-1-IG-150UL
This item is not returnable.
View return policy
Description
The GPD2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human GPD2. This antibody recognizes human, rat, mouse antigen. The GPD2 antibody has been validated for the following applications: WB, ELISA analysis.Specifications
| GPD2 | |
| Monoclonal | |
| 500 μg/mL | |
| GDH2; GPDM; mGPDH | |
| Recombinant Protein | |
| 150 μL | |
| Primary | |
| Human, Rat, Mouse | |
| Monoclonal Antibody | |
| IgG1 |
| Western Blot, ELISA | |
| 1C12E2 | |
| Unconjugated | |
| Mouse | |
| Protein G purification | |
| RUO | |
| 2820 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction