missing translation for 'onlineSavingsMsg'
Learn More
Learn More
gp96/HSP90B1/GRP94 Antibody (CL2647) [mFluor Violet 610 SE], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-42379MFV610
This item is not returnable.
View return policy
Description
gp96/HSP90B1/GRP94 Monoclonal antibody specifically detects gp96/HSP90B1/GRP94 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),KnockDown,Immunocytochemistry/ Immunofluorescence
Specifications
| gp96/HSP90B1/GRP94 | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| 94 kDa glucose-regulated protein, endothelial cell (HBMEC) glycoprotein, glucose regulated protein, 94 kDa, GP96ECGP, GRP-94, GRP94gp96 homolog, Heat shock protein 90 kDa beta member 1, heat shock protein 90kDa beta (Grp94), member 1, TRA1endoplasmin, tumor rejection antigen (gp96) 1, Tumor rejection antigen 1, Tumor rejection antigen-1 (gp96) | |
| This antibody was developed using a recombinant protein derived from P14625, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Adaptive Immunity, B Cell Development and Differentiation Markers, Cancer, Cellular Markers, Cytokine Research, ER Markers, Golgi Apparatus Markers, Hypoxia, Immunology, Membrane Trafficking and Chaperones, Mesenchymal Stem Cell Markers, Myeloid derived Suppressor Cell, Phospho Specific, Signal Transduction, Stem Cell Lines, Stem Cell Markers | |
| 7184 | |
| Store at 4⪚C in the dark. | |
| IgG2b |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), KnockDown, Immunocytochemistry/Immunofluorescence | |
| CL2647 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction