missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PYCR1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-38355-100ul
This item is not returnable.
View return policy
Description
PYCR1 Polyclonal antibody specifically detects PYCR1 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| PYCR1 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA | |
| ARCL2B, EC 1.5.1.2, mitochondrial pyrroline-5-carboxylate reductase 1, P5C, P5C reductase 1, P5CR, P5CR 1, PIG45, PP222, PRO3, proliferation-inducing protein 45, PYCR, pyrroline-5-carboxylate reductase 1, pyrroline-5-carboxylate reductase 1, mitochondrial | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-319 of human PYCR1 (NP_008838.2).,, Sequence:, MSVGFIGAGQLAFALAKGFTAAGVLAAHKIMASSPDMDLATVSALRKMGVKLTPHNKETVQHSDVLFLAVKPHIIPFILDEIGADIEDRHIVVSCAAGVTISSIEKKLSAFRPAPRVIRCMTNTPVVVREGATVYATGTHAQVEDGRLMEQLLSSVGFCTEVEEDLIDAVTGLSGSGPAYAFTALDALADGGVKMGLPRRLAVRLGAQALLGAAKMLLHSEQHPGQLKDNVSSPGGATIHALHVLESGGFRSLLINAVEASCIRTRELQSMADQEQVSPAAIKKTILDKVKLDSPAGTALSPSGHTKLLPRSLAPAGKD | |
| 100 μL | |
| metabolism | |
| 5831 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction