Learn More
Gosselin™ Round HDPE 100 mL Bottles
Product Code. p-8002228
Click to view available options
Closure Type:
Double Screw Cap
Screw Cap with Wad
Single Screw Cap
Closure Material:
HDPE
LDPE
Polyethylene
Tresylene
Closure Color:
Blue
Red
Sterility:
Non-sterile
Sterile
Unit Size:
Pack of 250
Pack of 335
Product Code. 11793293
Brand: Gosselin™ LR10004
Only null left
Add to basket
Add to basket
Provides excellent storage for liquids and solids in the laboratory. Gosselin™ Round HDPE 100mL Bottles come with either a screw cap with inner lip, with separate seal or with wad.
  • Large neck
  • Break resistant
  • Sterile or aseptic by irradiation
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 100 mL
Color Natural
Material HDPE
Length (Metric) 51mm
Quantity 335/PK
Sterility Non-sterile
Width (Metric) 51mm
Closure Color Red
Certifications/Compliance EC N°1935/2004
Closure Included Yes
Shape Round
Height (Metric) 80 mm
Closure Material LDPE
Diameter (Metric) 37 mm
Closure Type Single Screw Cap