missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goosecoid Antibody (SR1466), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22090-100ul
This item is not returnable.
View return policy
Description
Goosecoid Monoclonal antibody specifically detects Goosecoid in Human samples. It is validated for Western Blot, Immunoprecipitation
Specifications
| Goosecoid | |
| Monoclonal | |
| Unconjugated | |
| PBS (pH 7.4), 150 mM NaCl, 50% Glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunoprecipitation | |
| SR1466 | |
| Western Blot 1:500-1:2000, Immunoprecipitation 1:50 | |
| goosecoid homeobox, homeobox protein goosecoid | |
| A synthesized peptide derived from human Goosecoid (Uniprot #: P56915) | |
| 100 μg | |
| Stem Cells | |
| 145258 | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction