missing translation for 'onlineSavingsMsg'
Learn More

Goat IgG (H+L) Antibody, R&D Systems™

Product Code. 18078111
Change view
Click to view available options
Quantity:
0.5 mg
Unit Size:
500µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18078111 0.5 mg 500µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18078111 Supplier R&D Systems Supplier No. R401CABS
  • ONLINE EXCLUSIVE PRICE
    181.00€
    171.15€ / 500µg
    Save 9.85€
    5% Off
 Restricted Item
Estimated Shipment: 12-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

IgG Polyclonal specifically detects IgG in Goat samples. It is validated for Secondary, Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IgG
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot, ELISA
Formulation Lyophilized from a 0.2 μm filtered solution in NaH2PO4, NaCl and Sodium Azide with Trehalose. with Sodium Azide
Gene Alias Immunoglobulin G, ImmunoglobulinG
Gene Symbols IGHG1
Host Species Rabbit
Immunogen Goat IgG
Purification Method Antigen Affinity-purified
Quantity 0.5 mg
Regulatory Status RUO
Primary or Secondary Secondary
Gene ID (Entrez) 3500
Test Specificity This antibody reacts with heavy chains on goat IgG, and with light chains on all goat immunoglobulins. No reactivity is observed to non-immunoglobulin goat serum proteins, human, mouse, or rat IgG by IEP.
Reconstitution Reconstitute with sterile PBS. If 0.5 mL of PBS is used, the antibody concentration will be 1 mg/mL.
Target Species Goat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 6 months, -20°C to -70°C under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.