missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat IgG Fraction To Rabbit Immunoglobulins, HRP, Polyclonal Secondary Antibody, MP Biomedicals
Brand: MP Biomedicals 0855682
This item is not returnable.
View return policy
Description
Host Species: Goat
Species Reactivity: Rb
Enzyme Immunoassay, Immunohistochemistry, Western Blotting, Electron Microscopy
Specifications
| IgG | |
| HRP | |
| 2 mL | |
| Secondary | |
| Lyophilized |
| Polyclonal | |
| Goat | |
| RUO | |
| Rabbit |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction