missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat F(ab')2 anti-Mouse IgG1 Heavy Chain Secondary Antibody (Pre-adsorbed), Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP2-68546
This item is not returnable.
View return policy
Description
IgG1 Heavy Chain Polyclonal antibody specifically detects IgG1 Heavy Chain in Mouse samples. It is validated for Western Blot, ELISA, Immunofluorescence
Specifications
| IgG1 Heavy Chain | |
| Polyclonal | |
| Unconjugated | |
| Goat | |
| Chromatography | |
| RUO | |
| Secondary | |
| Store at 4°C. | |
| IgG |
| Western Blot, ELISA, Immunofluorescence | |
| 0.5 mg/mL | |
| Borate buffered saline | |
| Human kappa chains | |
| 0.5 mg | |
| Immunology | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction