missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat F(ab')2 anti-Human IgG Heavy Chain Secondary Antibody (Pre-adsorbed), Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP2-68543
This item is not returnable.
View return policy
Description
IgG Heavy Chain Polyclonal antibody specifically detects IgG Heavy Chain in Human samples. It is validated for Western Blot, ELISA, Immunofluorescence
Specifications
| IgG Heavy Chain | |
| Polyclonal | |
| Unconjugated | |
| Immunoglobulin G, ImmunoglobulinG | |
| Human IgG | |
| 0.5 mg | |
| Immunology | |
| Human | |
| Purified |
| Western Blot, ELISA, Immunofluorescence | |
| 0.5 mg/mL | |
| Borate buffered saline | |
| Goat | |
| Chromatography | |
| RUO | |
| Secondary | |
| Store at 4°C. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction