missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat F(ab')2 anti-Human IgG F(ab')2 Secondary Antibody, DyLight 649 (Pre-adsorbed), Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP1-51902
This item is not returnable.
View return policy
Description
IgG F(ab')2 Polyclonal antibody specifically detects IgG F(ab')2 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin).
Specifications
| IgG F(ab')2 | |
| Polyclonal | |
| DyLight 649 | |
| immunoglobulin heavy constant gamma 1 (G1m marker) | |
| Goat | |
| Affinity Purified | |
| RUO | |
| Secondary | |
| Antiserum was solid phase adsorbed to ensure specificity. Antiserum was cross adsorbed using mouse and rat immunosorbents to remove cross reactive antibodies. The antibody to human F(ab')2 was isolated by affinity chromatography using antigen coupled to agarose beads. F(ab')2 fragments were generated using a pepsin digestion. Fc fragments and whole IgG molecules have been removed. Fragments were conjugated to DyLight 649. Antibody concentration was determined by extinction coefficient: absorbance at 280 nm of 1.4 equals 1.0 mg of IgG. By immunoelectrophoresis and ELISA this antibody reacts specifically with human F(ab')2. No antibody was detected against non-immunoglobulin serum proteins. Less than 1% cross reactivity to mouse and rat IgG was detected. This antibody may cross react with F(ab')2 from other species. | |
| Store at 4C. Do not freeze. |
| Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 0.5 mg/ml | |
| Flow Cytometry 1:50 - 1:200, Immunohistochemistry 1:50 - 1:500, Immunocytochemistry/Immunofluorescence 1:50 - 1:500, Immunohistochemistry-Paraffin 1:50 - 1:500 | |
| IGHG1 | |
| Human IgG-F(ab')2 Fragment | |
| 1 mL | |
| Epitope Tags | |
| 3500 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction