missing translation for 'onlineSavingsMsg'
Learn More

Goat Anti-Syrian Hamster IgG H&L (Texas Red ™) preadsorbed, Abcam

Product Code. 32118836
Change view
Click to view available options
Quantity:
500 μg
Unit Size:
500µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32118836 500 μg 500µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 32118836 Supplier Abcam Supplier No. ab7144500ug
 Restricted Item
Estimated Shipment: 02-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Goat Polyclonal Antibody

Goat Anti-Syrian Hamster IgG H&L (Texas Red ™) preadsorbed

The product is subject to the following: Abcam Restricted Use Statement

.

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Antigen Syrian hamster IgG
Applications ELISA, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (FFPE), Immunohistochemistry (Frozen), Immunomicroscopy
Classification Polyclonal
Conjugate Texas Red
Formulation pH: 7.2, Preservative: 0.01% Sodium azide, Constituents: 1% BSA, 0.88% Sodium chloride, 0.424% Potassium phosphate solution
Host Species Goat
Immunogen Syrian hamster IgG
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Hamster
Content And Storage -20°C
Product Type Primary Antibody
Form Lyophilized
Isotype IgG
Shipping Condition Blue Ice
Target Molecule Syrian hamster IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.