missing translation for 'onlineSavingsMsg'
Learn More

Goat Anti-Rhesus monkey IgG H&L (PE), Abcam

Product Code. 32116991
Change view
Click to view available options
Quantity:
250 μg
Unit Size:
250µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32116991 250 μg 250µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 32116991 Supplier Abcam Supplier No. ab125852250ug
 Restricted Item
Estimated Shipment: 08-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Goat Polyclonal Antibody

Suitable for WB, Dot, ELISA, ICC/IF, IHC-P.

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Antigen Rhesus monkey IgG
Applications Dot Blot, ELISA, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (FFPE), Western Blot
Classification Polyclonal
Conjugate PE
Formulation pH: 7.3
Host Species Goat
Immunogen Rhesus monkey IgG
Purification Method Affinity purification
Quantity 250 μg
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Not Available
Content And Storage 4°C
Product Type Primary Antibody
Form Liquid
Isotype IgG
Shipping Condition Blue Ice
Target Molecule Rhesus monkey IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.