missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Rat IgG, Whole Molecule Polyclonal Secondary Antibody, HRP, MP Biomedicals
Shop All MP Biomedicals ProductsDescription
Host Species: Goat
Species Reactivity: Rat
Specifications
Specifications
| Antigen | IgG, Whole Molecule |
| Classification | Polyclonal |
| Conjugate | HRP |
| Host Species | Goat |
| Quantity | 2 mL |
| Primary or Secondary | Secondary |
| Target Species | Rat |
| Form | Lyophilized |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?