missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Rat IgG Fc Secondary Antibody, R-Phycoerythrin (Pre-adsorbed), Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP2-61920PE
This item is not returnable.
View return policy
Description
IgG Fc Polyclonal antibody specifically detects IgG Fc in Rat samples. It is validated for Flow Cytometry, Immunofluorescence.
Specifications
| IgG Fc | |
| Polyclonal | |
| R-PE | |
| PBS containing 0.2% BSA with 0.09% Sodium Azide | |
| Rat IgG-Fc Fragment | |
| 1 mL | |
| Secondary | |
| Rat | |
| IgG |
| Flow Cytometry, Immunofluorescence | |
| 0.5 mg/mL | |
| Flow Cytometry 1:50 - 1:200, Immunofluorescence 1:50 - 1:200 | |
| Goat | |
| Affinity Purified | |
| Adaptive Immunity | |
| Antiserum was solid phase adsorbed to ensure class specificity. Antiserum was cross adsorbed using human and mouse immunosorbents to remove cross reactive antibodies. The antibody to rat IgG was isolated by affinity chromatography using antigen coupled to agarose beads. F(ab')2 fragments were generated using a pepsin digestion. Fc fragments and whole IgG molecules have been removed. Fragments were conjugated to R-phycoerythrin (RPE). Antibody concentration was determined by extinction coefficient: absorbance at 280 nm of 1.4 equals 1.0 mg of IgG. By immunoelectrophoresis and ELISA this antibody reacts specifically with rat IgG. Cross reactivity with IgA and IgM is negligible. No antibody was detected against non-immunoglobulin serum proteins. Less than 1% cross reactivity to human and mouse IgG was detected. This antibody may cross react with IgG from other species. | |
| Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction