missing translation for 'onlineSavingsMsg'
Learn More

Goat anti-Rabbit IgG (H+L) Secondary Antibody, HRP (Pre-adsorbed), Novus Biologicals™

Product Code. 18291685 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
1000 μg
Unit Size:
1000µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18291685 1000 μg 1000µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18291685 Supplier Novus Biologicals Supplier No. NBP1753251000UG
  • ONLINE EXCLUSIVE PRICE
    338.00€
    319.20€ / 1000µg
    Save 18.80€
    5% Off
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Goat Polyclonal Antibody has been used in 2 publications

IgG (H+L) Polyclonal specifically detects IgG (H+L) in Rabbit samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IgG (H+L)
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate HRP
Dilution Western Blot 1:200-1:5000, ELISA 1:200-1:5000, Immunohistochemistry, Immunohistochemistry-Paraffin 1:500-1:5000
Formulation 10 mM Sodium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 1 % (w/v) BSA, Protease/IgG free
Host Species Goat
Immunogen Purified Rabbit IgG (H&L)
Quantity 1000 μg
Test Specificity Based on IEP, this antibody reacts with heavy gamma chains on rabbit IgG and light chains on all rabbit immunoglobulins
Reconstitution Rehydrate with 1.1 mL of deionized water and let stand 30 minutes at room temperature to dissolve. (Product has been overfilled to ensure complete recovery.) Centrifuge to remove any particulates. Prepare fresh working dilution daily.
Target Species Rabbit
Content And Storage Store lyophilized antibody at 4°C. Mix reconstituted liquid with an equal volume of glycerol, aliquot contents and freeze at -20°C or below for long term storage.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.