missing translation for 'onlineSavingsMsg'
Learn More

Goat anti-Rabbit IgG Fraction, Whole Molecule Polyclonal Secondary Antibody, HRP, MP Biomedicals

Product Code. 11430085 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
2 mL
Unit Size:
2mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11430085 2 mL 2mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11430085 Supplier MP Biomedicals Supplier No. 0855676
  • ONLINE EXCLUSIVE PRICE
    545.00€
    518.70€ / 2mL
    Save 26.30€
    4% Off
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Host Species: Goat
Isotype: IgG
Species Reactivity: Rabbit

Specifications

Antigen IgG, Whole Molecule
Classification Polyclonal
Conjugate HRP
Host Species Goat
Quantity 2 mL
Primary or Secondary Secondary
Target Species Rabbit
Form Purified
Isotype IgG
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.