missing translation for 'onlineSavingsMsg'
Learn More

Goat anti-Rabbit IgA Secondary Antibody, PerCP, Novus Biologicals™

Product Code. 30227007 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30227007 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30227007 Supplier Novus Biologicals Supplier No. NBP268338PCP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody

IgA Polyclonal antibody specifically detects IgA in Rabbit samples. It is validated for ELISA, Immunodiffusion
TRUSTED_SUSTAINABILITY

Specifications

Antigen IgA
Applications ELISA, Immunodiffusion
Classification Polyclonal
Conjugate PerCP
Formulation PBS
Gene Alias Immunoglobulin A
Host Species Goat
Immunogen Purified rabbit IgA
Purification Method Affinity purified
Quantity 1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Immunology
Primary or Secondary Secondary
Gene ID (Entrez) 3493
Target Species Rabbit
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.