missing translation for 'onlineSavingsMsg'
Learn More

Goat anti-Porcine IgG Fc Secondary Antibody, HRP, Novus Biologicals™

Product Code. 18201442 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18201442 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18201442 Supplier Novus Biologicals Supplier No. NB736
  • ONLINE EXCLUSIVE PRICE
    337.00€
    318.15€ / 1mL
    Save 18.85€
    5% Off
 Restricted Item
Estimated Shipment: 25-09-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Goat Polyclonal Antibody has been used in 2 publications

IgG Fc Polyclonal specifically detects IgG Fc in Pig samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IgG Fc
Applications ELISA, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate HRP
Dilution Western Blot 1:1000-1:30000, ELISA 1:10000-1:100000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:200 - 1:500, Immunohistochemistry-Paraffin 1:250-1:500
Host Species Goat
Immunogen Pig IgG-Fc Fragment
Purification Method Affinity Purified
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Secondary
Test Specificity By immunoelectrophoresis and ELISA this reacts specifically with pig IgG. This may cross react with IgG from other species.
Target Species Pig
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.