missing translation for 'onlineSavingsMsg'
Learn More

Goat Anti-Mouse IgM mu chain (DyLight™ 488), Abcam

Product Code. 32116539
Change view
Click to view available options
Quantity:
500 μg
Unit Size:
500µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32116539 500 μg 500µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 32116539 Supplier Abcam Supplier No. ab97007500ug
 Restricted Item
Estimated Shipment: 15-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Goat Polyclonal Antibody

Suitable for IHC-P, Flow Cyt, ICC/IF. Cited in 1 publication.

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Antigen Ighm
Applications Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (FFPE)
Cellular Localization Isoform 1: Secreted; During differentiation, B-lymphocytes switch from expression of membrane-bound IgM to secretion of IgM. Isoform 2: Cell membrane; Single-pass membrane protein
Classification Polyclonal
Conjugate DyLight 488
Formulation pH: 6.8 - 7.4, Preservative: 0.09% Sodium azide, Constituents: PBS, 0.2% BSA
Gene Ighm
Gene Accession No. P01872
Host Species Goat
Immunogen Ighm
Purification Method Affinity purification
Quantity 500 μg
Regulatory Status RUO
Research Discipline Immunology & Infectious Disease
Primary or Secondary Secondary
Target Species Mouse
Content And Storage 4°C
Product Type Primary Antibody
Form Liquid
Isotype IgG
Shipping Condition Blue Ice
Target Gene Ighm
Target Molecule Ighm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.