missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Mouse IgM Heavy Chain Secondary Antibody, FITC (Pre-adsorbed), Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP1-75192
This item is not returnable.
View return policy
Description
IgM Heavy Chain Polyclonal antibody specifically detects IgM Heavy Chain in Mouse samples. It is validated for Flow Cytometry, Immunomicroscopy.
Specifications
| IgM Heavy Chain | |
| Polyclonal | |
| FITC | |
| Constant region of heavy chain of IgM, Ig Mu Chain C Region BOT, Ig Mu Chain C Region GAL, Ig Mu Chain C Region OU, immunoglobulin heavy constant mu, Immunoglobulin mu chain | |
| Purified Mouse IgM, u chain | |
| 2 mg | |
| Immunology | |
| 3507 | |
| Mouse | |
| IgM |
| Flow Cytometry, Immunomicroscopy | |
| 1.0mg/mL | |
| Flow Cytometry 1:20-1:2000, Immunomicroscopy 1:20-1:2000 | |
| Goat | |
| Affinity purified | |
| RUO | |
| Secondary | |
| Based on IEP, this antibody reacts with heavy mu chains on mouse IgM. This antibody has been pre-adsorbed against human IgG and serum proteins. | |
| Store at 4C. After dilution, add an equal volume of glycerol to prevent loss of enzymatic activity and store at -20C. Avoid repeated freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction