missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Mouse IgG2b Secondary Antibody (Pre-adsorbed), Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP2-68512
This item is not returnable.
View return policy
Description
IgG2b Polyclonal antibody specifically detects IgG2b in Mouse samples. It is validated for ELISA
Specifications
| IgG2b | |
| Polyclonal | |
| Unconjugated | |
| Goat | |
| Affinity purified | |
| RUO | |
| Secondary | |
| Store at 4°C. | |
| IgG |
| ELISA | |
| 1 mg/mL | |
| Borate buffered saline | |
| Mouse IgG2b paraproteins | |
| 0.5 mg | |
| Neuronal Cell Markers, Neuroscience, Neurotransmission | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction