Learn More
Goat anti-Mouse IgG2b Secondary Antibody, DyLight 550, Novus Biologicals™
Product Code. 30227292 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.5 mL
Unit Size:
0.5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30227292 0.5 mL 0.5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30227292 Supplier Novus Biologicals Supplier No. NBP268512R
Goat Polyclonal Antibody
IgG2b Polyclonal antibody specifically detects IgG2b in Mouse samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY
Specifications
Antigen IgG2b
Applications ELISA
Classification Polyclonal
Conjugate DyLight 550
Formulation 50mM Sodium Borate
Host Species Goat
Immunogen Mouse IgG2b paraproteins
Purification Method Affinity purified
Quantity 0.5 mL
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG