missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Mouse IgG - H&L , Secondary Antibody, Abcam™
Description
Host Species: Goat
Isotype: IgG
Immunogen: mouse whole molecule
Applications:
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunomicroscopy, Western Blotting
Specifications
Specifications
| Antigen | Mouse IgG- H&L |
| Classification | Polyclonal |
| Concentration | 2mg/mL |
| Conjugate | FITC |
| Host Species | Goat |
| Quantity | 1 mg |
| Research Discipline | Immunology |
| Primary or Secondary | Secondary |
| Isotype | IgG |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?