missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Mouse IgA Secondary Antibody (Pre-adsorbed), Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP2-68523
364.65 EUR valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.
This item is not returnable.
View return policy
Description
IgA Polyclonal antibody specifically detects IgA in Mouse samples. It is validated for ELISA
Specifications
| IgA | |
| Polyclonal | |
| Unconjugated | |
| Immunoglobulin A | |
| Mouse IgA paraproteins | |
| 0.5 mg | |
| Adaptive Immunity, Immunology | |
| 3493 | |
| Store at 4°C. | |
| IgG |
| ELISA | |
| 1 mg/mL | |
| Borate buffered saline | |
| Goat | |
| Affinity purified | |
| RUO | |
| Secondary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction