missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Mouse IgA/IgG/IgM (H+L) Secondary Antibody, Alkaline Phosphatase (Pre-adsorbed), Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP1-72648
This item is not returnable.
View return policy
Description
IgA/IgG/IgM (H+L) Polyclonal specifically detects IgA/IgG/IgM (H+L) in Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry.
Specifications
| IgA/IgG/IgM (H+L) | |
| Polyclonal | |
| Alkaline Phosphatase (AP) | |
| Goat | |
| Affinity Purified | |
| RUO | |
| No reaction was observed against bovine, horse or human serum proteins. This reagent is suitable for the detection of all Mouse immunoglobulin isotypes, subclasses and chain combinations. | |
| Store at 4C. Do not freeze. |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| Western Blot 1:500 - 1:2500, ELISA 1:2000 - 1:10000, Immunohistochemistry 1:200 - 1:1000 | |
| Mouse IgG IgA and IgM whole molecule | |
| 0.5 mg | |
| Secondary | |
| Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction