missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Monkey IgG (H+L) Secondary Antibody, HRP, Novus Biologicals™
Goat Polyclonal Antibody has been used in 3 publications
Brand: Novus Biologicals NB7215
This item is not returnable.
View return policy
Description
IgG (H+L) Polyclonal specifically detects IgG (H+L) in Monkey samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| IgG (H+L) | |
| Polyclonal | |
| HRP | |
| Goat | |
| Affinity Purified | |
| RUO | |
| Secondary | |
| Monkey | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1.0 mg/mL | |
| Western Blot 1:5000- 1:50000, ELISA 1:10000- 1:100000, Immunohistochemistry 1:200 - 1:5000, Immunocytochemistry/Immunofluorescence 1:200 - 1:5000, Immunohistochemistry-Paraffin | |
| Monkey IgG-heavy and light chain | |
| 1 mL | |
| Adaptive Immunity, Immunology | |
| By immunoelectrophoresis and ELISA this antibody reacts specifically with monkey IgG and with light chains common to other monkey immunoglobulins. Species detected are Rhesus, Baboon, Carb Green, Cyno, Chimp and African Green. Reaction to human was also detected. No antibody was detected against non-immunoglobulin serum proteins. This antibody may cross react with IgG from other species. | |
| Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction