missing translation for 'onlineSavingsMsg'
Learn More

Goat Anti-Human IgG (Fab')2 (Alkaline Phosphatase), Abcam

Product Code. 32117265
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32117265 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 32117265 Supplier Abcam Supplier No. ab87408100ug
 Restricted Item
Estimated Shipment: 02-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Goat Polyclonal Antibody

Goat Anti-Human IgG (Fab')2 (Alkaline Phosphatase)

The product is subject to the following: Abcam Restricted Use Statement

.

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Antigen IGHG1
Applications ELISA, Western Blot
Cellular Localization Isoform 1: Secreted; Isoform 2: Cell membrane; Single-pass membrane protein
Classification Polyclonal
Conjugate Alkaline Phosphatase (AP)
Formulation Preservative: 0.05% Sodium azide, Constituents: Tris buffered saline, 1.5% BSA
Gene IGHG1
Gene Accession No. P01857
Host Species Goat
Immunogen IGHG1
Purification Method Affinity purification
Quantity 100 μg
Regulatory Status RUO
Research Discipline Immunology & Infectious Disease
Primary or Secondary Secondary
Target Species Human
Content And Storage 4°C
Product Type Primary Antibody
Form Liquid
Isotype IgG
Shipping Condition Blue Ice
Target Gene IGHG1
Target Molecule IGHG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.