missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Human IgG F(ab) Secondary Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP2-68537
This item is not returnable.
View return policy
Description
IgG F(ab) Polyclonal antibody specifically detects IgG F(ab) in Human samples. It is validated for ELISA
Specifications
| IgG F(ab) | |
| Polyclonal | |
| Unconjugated | |
| Goat | |
| Affinity purified | |
| RUO | |
| Human | |
| Purified |
| ELISA | |
| 0.5 mg/mL | |
| Borate buffered saline | |
| Human IgG | |
| 0.5 mg | |
| Secondary | |
| Store at 4°C. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction