missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Hamster IgG (H+L) Secondary Antibody, FITC (Pre-adsorbed), Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP1-73711
This item is not returnable.
View return policy
Description
IgG (H+L) Polyclonal specifically detects IgG (H+L) in Hamster samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Fluorophore-linked immunosorbent assay.
Specifications
| IgG (H+L) | |
| Polyclonal | |
| Fluorescein | |
| Goat | |
| Affinity Purified | |
| RUO | |
| Secondary | |
| Reconstitute with 1.0 ml deionized water (or equivalent). | |
| Store at 4C in the dark. |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
| LYOPH | |
| Flow Cytometry 1:1:10 - 1:1000, Immunocytochemistry/Immunofluorescence 1:500-1:2500, Fluorophore-linked immunosorbent assay 1:100 - 1:2000 | |
| Armenian and Golden Syrian Hamster IgG, whole molecule | |
| 1 mg | |
| Adaptive Immunity, Immunology | |
| This antibody was pre-adsorbed against Mouse and Rat Serum Proteins. No reaction was observed against Mouse or Rat Serum Proteins. | |
| Hamster | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction