missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Hamster IgG (H+L) Secondary Antibody, Alkaline Phosphatase (Pre-adsorbed), Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP1-72682
This item is not returnable.
View return policy
Description
IgG (H+L) Polyclonal specifically detects IgG (H+L) in Hamster samples. It is validated for Western Blot, ELISA, Immunohistochemistry.
Specifications
| IgG (H+L) | |
| Polyclonal | |
| Western Blot 1:500 - 1:3000, ELISA 1:2000 - 1:10000, Immunohistochemistry 1:200-1:1000 | |
| Armenian and Golden Syrian Hamster IgG, whole molecule | |
| 1 mg | |
| Adaptive Immunity, Immunology | |
| This antibody was pre-adsorbed against Mouse and Rat Serum Proteins. No reaction was observed against Mouse or Rat Serum Proteins. | |
| Store at 4C. Do not freeze. |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Alkaline Phosphatase (AP) | |
| Goat | |
| Affinity Purified | |
| RUO | |
| Secondary | |
| Hamster | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction