Learn More
CYP26B1, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of CYP26B1.
Brand: Abnova PAB7139.100ug
Description
This gene encodes a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases that catalyze many reactions involved in drug metabolism and the synthesis of cholesterol, steroids and other lipids. The enzyme encoded by this gene is involved in the specific inactivation of all-trans-retinoic acid to hydroxylated forms, such as 4-oxo-, 4-OH-, and 18-OH-all-trans-retinoic acid. [provided by RefSeq
Sequence: C-SQARSEDKDGRFHSpecifications
| CYP26B1 | |
| Polyclonal | |
| Unconjugated | |
| ELISA (1:128000) The optimal working dilution should be determined by the end user. | |
| CYP26B1 | |
| CYP26A2/DKFZp686G0638/MGC129613/P450RAI-2 | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 56603 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Immunohistochemistry (PFA fixed) | |
| 0.5 mg/mL | |
| Goat polyclonal antibody raised against synthetic peptide of CYP26B1. | |
| In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
| NP_063938.1 | |
| CYP26B1 | |
| A synthetic peptide corresponding to human CYP26B1. | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only