missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CLU Goat anti-Human, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against native CLU.
Brand: Abnova PAB9973.500ug
This item is not returnable.
View return policy
Specifications
| CLU | |
| Polyclonal | |
| Goat polyclonal antibody raised against native CLU. | |
| In 100mM Na2B4O7, 75mM NaCl, 5mM EDTA, pH 8.0 (0.01% sodium azide) | |
| AAG4/APOJ/CLI/KUB1/MGC24903/SGP-2/SGP2/SP-40/TRPM-2/TRPM2 | |
| Goat | |
| 500 μg | |
| Primary | |
| This antibody reacts with human apoLipoprotein J and has negligible cross-reactivity with Type A-I, A-II, B, C-I, C-II, C-III and E apoLipoproteins. Non-specific cross reaction of anti-apoLipoprotein antibodies with other human serum proteins is negligible. | |
| Store at 4°C. For long term storage store with 50% glycerol at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot | |
| Unconjugated | |
| ELISA (1:5000-1:10000) Western Blot (1:5000-1:10000) Immunohistochemistry (1:50-1:200) The optimal working dilution should be determined by the end user. | |
| CLU | |
| CLU | |
| Native purified human CLU. | |
| RUO | |
| 1191 | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction